• USA Home
  • 81132 - Poly(acrylic acid sodium salt)

EMAIL THIS PAGE TO A FRIEND
81132 Aldrich

Poly(acrylic acid sodium salt)

5,100

Synonym: Poly(sodium acrylate), Sodium polyacrylate

Purchase

Properties

Related Categories Hydrophilic Polymers, Materials Science, Poly(acrylic acid) (PAA) Salts and Solutions, Poly(acrylic acid) (PAA) and Copolymers, Poly(acrylic acid), Polymethacrylate and Other Acrylic Polymers,
form   crystalline
mol wt   Mw 5550
impurities   ≤15% water
pH   6.0-9.0 (10 mg/mL in H2O)
cation traces   Na: ~19%

Price and Availability


Laboratory Gloves

Webinar: Recent Developments in RAFT Agents

The Future of Bioimaging
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
WD6826000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?