• USA Home
  • 81185 - Polyethylene glycol 600 bound to polystyrene - 1% DVB

EMAIL THIS PAGE TO A FRIEND
81185 Aldrich

Polyethylene glycol 600 bound to polystyrene - 1% DVB

extent of labeling: 0.35 mmol/g OH loading

Synonym: Polyethylene glycol 600 monobenzyl ether polymer-bound

  • Linear Formula PS-CH2O(CH2CH2O)nH n~13

  •  MDL number MFCD00147995

Purchase

Properties

Related Categories Chemical Synthesis, PEG Resins, Solid Supported Synthesis, Specialty Synthesis
extent of labeling   0.35 mmol/g OH loading

Description

Other Notes

Immobilized polyethylene glycols are highly active and stable triphase catalysts for dehydro-halogenation reactions in two-phase systems

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?