EMAIL THIS PAGE TO A FRIEND
81553 Aldrich

Polystyrene AM-NH2

macrobeads, extent of labeling: 0.8-1.4 mmol/g N loading

Synonym: (Aminomethyl)polystyrene, macrobeads

Purchase

Properties

Related Categories Base Resins for SPPS, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Polymer Supports for SPPS More...
extent of labeling   0.8-1.4 mmol/g N loading
matrix   crosslinked with 1% DVB
particle size   160-200 μm
storage temp.   2-8°C

Description

Packaging

10 g in poly bottle

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Aminoalkylated Resins

Our (aminomethyl)polystyrene resins are prepared by aminomethylation of unsubstituted polystyrene in order to ensure chemically homogeneous supports, free of any reactive chloromethyl groups. Aminoal...
Chemfiles Volume 3 Article 4
Keywords: Aminomethylations, Immobilization, Peptide synthesis, Solid phase peptide synthesis, Solid supported organic synthesis

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?