• USA Home
  • 81562 - Polystyrene A-S-Trityl

EMAIL THIS PAGE TO A FRIEND
81562 Aldrich

Polystyrene A-S-Trityl

extent of labeling: 0.6-0.8 mmol/g loading

Synonym: 2-[3-(Tritylthio)propionylamino]ethyl-polystyrene

Purchase

Properties

Related Categories Chemical Synthesis, Polystyrene Resins, Solid Supported Synthesis, Specialty Synthesis
extent of labeling   0.6-0.8 mmol/g loading
matrix   crosslinked with 1% DVB
particle size   160-200 μm
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?