• USA Home
  • 82468 - Triphenylpropylphosphonium bromide

EMAIL THIS PAGE TO A FRIEND
82468 Aldrich

Triphenylpropylphosphonium bromide

≥98.0% (AT)

Synonym: NSC 50539, Propyltriphenylphosphonium bromide

  • CAS Number 6228-47-3

  • Linear Formula (C6H5)3P(Br)CH2CH2CH3

  • Molecular Weight 385.28

  •  Beilstein Registry Number 3599726

  •  EC Number 228-330-2

  •  MDL number MFCD00011843

  •  PubChem Substance ID 24887975

Purchase

Properties

Related Categories C-C Bond Formation, Chemical Synthesis, Olefination, Synthetic Reagents, Wittig Reagents More...
InChI Key   XMQSELBBYSAURN-UHFFFAOYSA-M
assay   ≥98.0% (AT)

Description

Application

Catalyst for the carboxylation of styrene oxide

Reactant involved in:
• Epoxidation of arylalkenes
• Boration of allylic epoxides
• Wittig reactions
• Selective dimerization of α-olefins
• Synthesis of indoles

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?