EMAIL THIS PAGE TO A FRIEND
850454 Aldrich

Progesterone

98%

Synonym: 4-Pregnene-3,20-dione

  • CAS Number 57-83-0

  • Empirical Formula (Hill Notation) C21H30O2

  • Molecular Weight 314.46

  •  Beilstein Registry Number 1915950

  •  EC Number 200-350-6

  •  MDL number MFCD00003658

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   RJKFOVLPORLFTN-LEKSSAKUSA-N
assay   98%
optical activity   [α]23/D +182°, c = 2 in dioxane
mp   128-132 °C(lit.)
Gene Information   human ... ESR1(2099), PGR(5241)

Description

Other Notes

Steroid hormone produced by the corpus luteum.

Biochem/physiol Actions

Induces maturation and secretory activity of the uterine endothelium; suppresses ovulation. Progesterone is implicated in the etiology of breast cancer.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
TW0175000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P0130 Progesterone, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?