• USA Home
  • 854204 - Protease from Streptomyces griseus

EMAIL THIS PAGE TO A FRIEND
854204 Aldrich

Protease from Streptomyces griseus

Purchase

Properties

activity   10 chymotrypsin units/mg dry wt.
  140 trypsin units/mg dry wt.

Description

Other Notes

Contains α-amylase

Physical form

A salt-free powder

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?