• USA Home
  • 855030 - β-D-Ribopyranose 1,2,3,4-tetraacetate

EMAIL THIS PAGE TO A FRIEND
855030 Aldrich

β-D-Ribopyranose 1,2,3,4-tetraacetate

99%

Synonym: 1,2,3,4-Tetra-O-acetyl-β-D-ribopyranose

Purchase

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology, Chemical Synthesis
InChI Key   MJOQJPYNENPSSS-LPWJVIDDSA-N
assay   99%
optical activity   [α]22/D −55.4°, c = 3 in methanol
mp   110-112 °C(lit.)

Description

Application

Intermediate in the preparation of nucleosides.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?