• USA Home
  • 855626 - N-Acetyl-2-fluoro-DL-phenylalanine

EMAIL THIS PAGE TO A FRIEND
855626 Aldrich

N-Acetyl-2-fluoro-DL-phenylalanine

98%

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylalanine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   ZKZPFAQDTBSPJH-UHFFFAOYSA-N
assay   98%
mp   153-155 °C(lit.)

Description

Packaging

5 g in glass bottle

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?