• USA Home
  • 856592 - β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate

EMAIL THIS PAGE TO A FRIEND
856592 Aldrich

β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate

Synonym: Triphosphopyridine nucleotide sodium salt hydrate

  • CAS Number 698999-85-8

  • Empirical Formula (Hill Notation) C21H27N7NaO17P3 · xH2O

  • Molecular Weight 765.39 (anhydrous basis)

  •  MDL number MFCD00150480

  •  PubChem Substance ID 329769396

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

InChI Key   CYQLUFJYVTUUFN-WUEGHLCSSA-M
impurities   <5% acetone
mp   175-178 °C (dec.)(lit.)

Description

Biochem/physiol Actions

Electron acceptor

Other Notes

This is the common form of NADP. Do not confuse with 3′−NADP or α-NADP.

Packaging

Packaged by solid weight.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1
Protocols & Articles

Related Content

Indoles, Purines, and Their Isosteres

Substituted indoles and purines have frequently been referred to as “privileged structures” since they are capable of binding to multiple receptors with high affinity, and thus have applications acro...
Keywords: Building blocks, Medicinal chemistry

Related Products

replaced by

Product #

Description

Add to Cart

N0505 β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?