• USA Home
  • 856762 - L-Alanine tert-butyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
856762 Aldrich

L-Alanine tert-butyl ester hydrochloride

  • CAS Number 13404-22-3

  • Linear Formula CH3CH(NH2)COOC(CH3)3 · HCl

  • Molecular Weight 181.66

  •  Beilstein Registry Number 3562140

  •  EC Number 236-497-8

Purchase

Properties

InChI Key   WIQIWPPQGWGVHD-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?