EMAIL THIS PAGE TO A FRIEND
856819 Aldrich

Ala-Ala-Ala

Synonym: Tri-L-alanine

  • CAS Number 5874-90-8

  • Empirical Formula (Hill Notation) C9H17N3O4

  • Molecular Weight 231.25

  •  MDL number MFCD00025557

Purchase

Properties

InChI Key   BYXHQQCXAJARLQ-ZLUOBGJFSA-N

Description

Amino Acid Sequence

Ala-Ala-Ala

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?