EMAIL THIS PAGE TO A FRIEND
856983 Aldrich

Ala-Phe

  • CAS Number 3061-90-3

  • Empirical Formula (Hill Notation) C12H16N2O3

  • Molecular Weight 236.27

  •  MDL number MFCD00066031

  •  PubChem Substance ID 329769404

Purchase

Properties

InChI Key   OMNVYXHOSHNURL-WPRPVWTQSA-N

Description

Amino Acid Sequence

Ala-Phe

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?