• USA Home
  • 857947 - Adenosine 5′-triphosphate monosodium salt

EMAIL THIS PAGE TO A FRIEND
857947 Aldrich

Adenosine 5′-triphosphate monosodium salt

  • CAS Number 15237-44-2

  • Empirical Formula (Hill Notation) C10H16N5NaO13P3

  • Molecular Weight 530.17

  •  PubChem Substance ID 329769415

Purchase

Properties

InChI Key   RMJPDRUNCDRUQC-MCDZGGTQSA-N

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?