• USA Home
  • 858048 - 6-(4-Carboxybutyl)mercaptopurine

EMAIL THIS PAGE TO A FRIEND
858048 Aldrich

6-(4-Carboxybutyl)mercaptopurine

Synonym: Buthiopurine

  • CAS Number 608-12-8

  • Empirical Formula (Hill Notation) C10H12N4O2S

  • Molecular Weight 252.29

  •  PubChem Substance ID 329769416

Purchase

Properties

InChI Key   FWYNOXVCJSRVQV-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?