• USA Home
  • 858900 - L-Aspartyl-L-phenylalanine methyl ester

EMAIL THIS PAGE TO A FRIEND
858900 Aldrich

L-Aspartyl-L-phenylalanine methyl ester

96%

Synonym: N-L-α-Aspartyl-L-phenylalanine methyl ester, Asp-Phe methyl ester, Aspartame

Purchase

Properties

InChI Key   IAOZJIPTCAWIRG-QWRGUYRKSA-N
assay   96%
mp   248-250 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
WM3407000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A5139 Asp-Phe methyl ester, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?