EMAIL THIS PAGE TO A FRIEND
859281 Aldrich

Gly-Asp dihydrate

Synonym: Glycyl-L-aspartic acid dihydrate

  • CAS Number 4685-12-5

  • Linear Formula NH2CH2CONHCH(COOH)CH2COOH · 2H2O

  • Molecular Weight 226.18

  •  Beilstein Registry Number 1727387

  •  MDL number MFCD00180728

  •  PubChem Substance ID 329769426

Purchase

Properties

InChI Key   SKIXLVGMUJDROR-DFWYDOINSA-N

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?