• USA Home
  • 860344 - 4-Nitrophenyl β-D-galactopyranoside hydrate

EMAIL THIS PAGE TO A FRIEND
860344 Aldrich

4-Nitrophenyl β-D-galactopyranoside hydrate

98%

Purchase

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology, Chemical Synthesis
InChI Key   KSEGJDPERYLUSV-YOGPMPDXSA-N
assay   98%
optical activity   [α]22/D −84.0°, c = 0.3 in H2O
mp   180-184 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

N1252 4-Nitrophenyl β-D-galacto­pyran­oside, ≥98% (enzymatic)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?