• USA Home
  • 861529 - bisBenzimide H 33342 trihydrochloride

EMAIL THIS PAGE TO A FRIEND
861529 Aldrich

bisBenzimide H 33342 trihydrochloride

Synonym: 2′-(4-Ethoxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5′-bi-1H-benzimidazole trihydrochloride, HOE 33342, Hoechst 33342, bisBenzimide

  • CAS Number 23491-52-3

  • Empirical Formula (Hill Notation) C27H28N6O · 3HCl · xH2O

  • Molecular Weight 561.93 (anhydrous basis)

  •  Beilstein Registry Number 1234011

  •  MDL number MFCD00012678

  • Popular Documents:  RAMAN FTIR (PDF)  

Purchase

Properties

InChI Key   FYEVKHPLBHLWHK-UHFFFAOYSA-N

Description

Application

Useful for staining DNA, chromosomes and nuclei. May be used for fluorescence microscopy or flow cytometry.
Excitation max. = 346 nm
Emission max. = 460 nm

Biochem/physiol Actions

Membrane-permeable, fluorescent DNA stains with low cytotoxicity that intercalate in A-T regions of DNA.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Indoles, Purines, and Their Isosteres

Substituted indoles and purines have frequently been referred to as “privileged structures” since they are capable of binding to multiple receptors with high affinity, and thus have applications acro...
Keywords: Building blocks, Medicinal chemistry

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?