• USA Home
  • 86332 - TentaGel HL-CH(OEt)2

EMAIL THIS PAGE TO A FRIEND
86332 Aldrich

TentaGel HL-CH(OEt)2

extent of labeling: ~0.40 mmol/g loading

Synonym: O-[2-(6,6-Diethoxyhexanoylamino)ethyl]polyethylene glycol polymer-bound

Purchase

Properties

Related Categories Chemical Synthesis, Solid Supported Synthesis, Specialty Synthesis, TentaGel Resins
extent of labeling   ~0.40 mmol/g loading
particle size   ~110 μm
storage temp.   2-8°C

Description

Legal Information

TentaGel is a trademark of Rapp Polymere GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?