EMAIL THIS PAGE TO A FRIEND
86363 Aldrich

TentaGel S-Br

extent of labeling: ~0.26 mmol/g loading

Synonym: O-(2-Bromoethyl)polyethylene glycol polymer-bound

Purchase

Properties

Related Categories Chemical Synthesis, Solid Supported Synthesis, Specialty Synthesis, TentaGel Resins
extent of labeling   ~0.26 mmol/g loading
particle size   ~90 μm
storage temp.   2-8°C

Description

Packaging

5 g in glass bottle

Legal Information

TentaGel is a trademark of Rapp Polymere GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Brominated Polystyrenes and Bromoalkylated Resins

Brominated polystyrene is a core matrix for the preparation of various functionalized polystyrene resins for SPPS and SPOS. The brominated polymer can be derivatized by Suzuki coupling reactions(1) o...
Chemfiles Volume 3 Article 4
Keywords: Acylations, Coupling reactions, Lithiations, Solid phase peptide synthesis, Solid supported organic synthesis, Suzuki coupling

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?