• USA Home
  • 86379 - TentaGel S PHB-Asp(t-Bu)Fmoc

EMAIL THIS PAGE TO A FRIEND
86379 Aldrich

TentaGel S PHB-Asp(t-Bu)Fmoc

extent of labeling: ~0.22 mmol/g protected amino acid loading

Synonym: Fmoc-L-aspartic acid 4-tert-butyl ester1-{4-[poly(ethylenoxy)]benzyl ester} polymer-bound

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Polymer Supports for SPPS, Pre-Loaded Resins,
extent of labeling   ~0.22 mmol/g protected amino acid loading
particle size   ~90 μm
storage temp.   2-8°C

Description

Legal Information

TentaGel is a trademark of Rapp Polymere GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Tentagel® S PHB Resins Preloaded with N-α-Fmoc-Protected Amino Acids

Preloaded TentaGel® S PHB resins are excellent acid-labile supports for Fmoc-/tBu SPPS of peptide acids. Due to the unique properties of the TentaGel® resin, synthesis can be performed both in a batc...
Chemfiles Volume 3 Article 4
Keywords: Epimerizations, Protocols, Solid phase peptide synthesis

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?