• USA Home
  • 86427 - TentaGel S Trt-Met-Fmoc

EMAIL THIS PAGE TO A FRIEND
86427 Aldrich

TentaGel S Trt-Met-Fmoc

extent of labeling: ~0.20 mmol/g protected amino acid loading

Synonym: Fmoc-L-methionine 4-[poly(oxyethylene)carbamoyl]trityl ester polymer-bound

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Polymer Supports for SPPS, Pre-Loaded Resins,
extent of labeling   ~0.20 mmol/g protected amino acid loading
particle size   ~90 μm
storage temp.   2-8°C

Description

Legal Information

TentaGel is a trademark of Rapp Polymere GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

TentaGel® S Trt Resins Preloaded with N-Terminal Free Amino Acids

Preloaded TentaGel® S Trt resin consists of Fmoc-amino acids attached to highly acid-labile 4-carboxytrityl resin.(1-3) TentaGel® S Trt is derived from TentaGel® S NH2 by acylation with 4-carboxytrit...
ChemFiles Volume 3 Article 4
Keywords: Acylations, Applications

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?