EMAIL THIS PAGE TO A FRIEND
86484 Aldrich

Terpinolene

purum, ≥95.0% (GC)

Synonym: 4-Isopropylidene-1-methylcyclohexene, p-Meth-1-en-8-yl-formate, p-Menth-1,4(8)-diene

  • CAS Number 586-62-9

  • Empirical Formula (Hill Notation) C10H16

  • Molecular Weight 136.23

  •  Beilstein Registry Number 1851203

  •  EC Number 209-578-0

  •  MDL number MFCD00049191

  •  PubChem Substance ID 329769466

Purchase

Properties

Related Categories Alkenes, Artemisia vulgaris, Boswellia carterii, Building Blocks, Cell Biology,
vapor density   ~4.7 (vs air)
vapor pressure   ~0.5 mmHg ( 20 °C)
InChI Key   MOYAFQVGZZPNRA-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.489(lit.)
  n20/D 1.490
bp   184-185 °C(lit.)
density   0.861 g/mL at 25 °C(lit.)
storage temp.   −20°C

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2541 3 / PGIII
WGK Germany 
2
RTECS 
WZ6870000
Flash Point(F) 
147.2 °F
Flash Point(C) 
64 °C
Protocols & Articles

Articles

GC Analysis of Cold Pressed Lemon Essential Oil on Equity®-5 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Cold Pressed Lemon Essential Oil on Equity®-5 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Size-exclusion chromatography

GC Analysis of Distilled Lime Essential Oil on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Distilled Lime Essential Oil on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Distilled Lime Essential Oil on Equity®-1 and Equity®-5

From our library of Articles, Sigma-Aldrich presents GC Analysis of Distilled Lime Essential Oil on Equity®-1 and Equity®-5
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Lemon Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Lemon Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Lemon Essential Oil on SLB®-IL59, 3 °C/min Oven Ramp

From our library of Articles, Sigma-Aldrich presents GC Analysis of Lemon Essential Oil on SLB®-IL59, 3 °C/min Oven Ramp
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Peppermint Essential Oil on SLB®-IL59

From our library of Articles, Sigma-Aldrich presents GC Analysis of Peppermint Essential Oil on SLB®-IL59
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Peppermint Essential Oil on SUPELCOWAX® 10

From our library of Articles, Sigma-Aldrich presents GC Analysis of Peppermint Essential Oil on SUPELCOWAX® 10
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Spearmint Essential Oil on SLB®-IL59

From our library of Articles, Sigma-Aldrich presents GC Analysis of Spearmint Essential Oil on SLB®-IL59
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Sweet Orange Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Sweet Orange Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Volatiles in Limoncello on SLB®-5ms after SPME using a 100 μm PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Volatiles in Limoncello on SLB®-5ms after SPME using a 100 μm PDMS Fiber
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Mass spectrometry, Purification, Sample preparations, Solid phase microextractions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?