EMAIL THIS PAGE TO A FRIEND
86557 Aldrich

H-Val-2-ClTrt resin

extent of labeling: ~1.0 mmol/g loading

Synonym: L-Valine 2-chlorotrityl ester polymer-bound

Purchase

Properties

Related Categories 2-Chlorotrityl Resin, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Polymer Supports for SPPS,
extent of labeling   ~1.0 mmol/g loading
particle size   75-150 μm
storage temp.   2-8°C

Description

Packaging

1 g in poly tube

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

2-Chlorotrityl Resins Preloaded with N-α-Fmoc-Protected Amino Acids

The mild cleavage conditions for highly acid-labile 2-chlorotrityl resin enables the preparation of fully protected peptide fragments for convergent synthesis and selective side-chain derivatization....
Chemfiles Volume 3 Article 4
Keywords: Coupling reactions, Derivatizations

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?