• USA Home
  • 86609 - Tetraethylammonium cyanate

EMAIL THIS PAGE TO A FRIEND
86609 Aldrich

Tetraethylammonium cyanate

technical

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   YVMHKCAQRHAEMF-UHFFFAOYSA-M
grade   technical
mp   ~270 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?