• USA Home
  • 86841 - Tetrabutylammonium succinimide

EMAIL THIS PAGE TO A FRIEND
86841 Aldrich

Tetrabutylammonium succinimide

≥97.0% (NT)

Synonym: Succinimide tetrabutylammonium salt

  • CAS Number 74830-30-1

  • Empirical Formula (Hill Notation) C20H40N2O2

  • Molecular Weight 340.54

  •  Beilstein Registry Number 4628898

  •  MDL number MFCD00210091

  •  PubChem Substance ID 57653196

Purchase

Properties

Related Categories Ammonium, Ammonium Salts, Chemical Synthesis, Ionic Liquids, Phase Transfer Catalysts,
InChI Key   OYSUQMQPIJNEBF-UHFFFAOYSA-M
assay   ≥97.0% (NT)
mp   ~75 °C

Description

Application

This lipophilic quaternary ammonium salt allows reactions with the succinimide anion to be run in aprotic, organic solvents

Packaging

5 g in glass bottle

Price and Availability


Laboratory Gloves

Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?