• USA Home
  • 87292 - Tetraheptylammonium chloride

EMAIL THIS PAGE TO A FRIEND
87292 Aldrich

Tetraheptylammonium chloride

≥98.0% (AT)

  • CAS Number 10247-90-2

  • Linear Formula [CH3(CH2)6]4N(Cl)

  • Molecular Weight 446.24

  •  Beilstein Registry Number 4167417

  •  EC Number 233-583-7

  •  MDL number MFCD00041983

  •  PubChem Substance ID 24888872

Purchase

Properties

Related Categories Ammonium, Chemical Synthesis, Ionic Liquids, Specialty Synthesis
InChI Key   VMJQVRWCDVLJSI-UHFFFAOYSA-M
assay   ≥98.0% (AT)
bp   175 °C(lit.)
mp   38-40 °C(lit.)
  38-41 °C

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?