• USA Home
  • 87304 - Tetrahexylammonium benzoate solution

EMAIL THIS PAGE TO A FRIEND
87304 Aldrich

Tetrahexylammonium benzoate solution

~75% in methanol

Synonym: THAB

  • CAS Number 16436-29-6

  • Linear Formula [CH3(CH2)5]4N(C6H5COO)

  • Molecular Weight 475.79

  •  Beilstein Registry Number 4084122

  •  MDL number MFCD00041982

  •  PubChem Substance ID 57653237

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   PSEQWFPWQRZBOO-UHFFFAOYSA-M
concentration   ~75% in methanol

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
3
Flash Point(F) 
53.6 °F
Flash Point(C) 
12 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?