• USA Home
  • 87406 - Chloro(chloromethyl)dimethylsilane

EMAIL THIS PAGE TO A FRIEND
87406 Aldrich

Chloro(chloromethyl)dimethylsilane

produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0% (GC)

Synonym: (Chloromethyl)dimethylchlorosilane, CMDMCS, Silan CMM2

  • CAS Number 1719-57-9

  • Linear Formula ClCH2Si(CH3)2Cl

  • Molecular Weight 143.09

  •  Beilstein Registry Number 1731933

  •  EC Number 217-006-6

  •  MDL number MFCD00000875

  •  PubChem Substance ID 24888896

Purchase

Properties

Related Categories Chemical Synthesis, Protecting and Derivatizing Reagents, Protection and Derivatization, Silicon-Based, Synthetic Reagents More...
grade   produced by Wacker Chemie AG, Burghausen, Germany
InChI Key   ITKVLPYNJQOCPW-UHFFFAOYSA-N
assay   ≥98.0% (GC)
refractive index   n20/D 1.437(lit.)
bp   114 °C/752 mmHg(lit.)
density   1.086 g/mL at 25 °C(lit.)

Description

Other Notes

prices for bulk quantities on request

Packaging

250 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 2985 8(3) / PGII
WGK Germany 
3
Flash Point(F) 
71.6 °F
Flash Point(C) 
22 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?