• USA Home
  • 87556 - 2-[2-(Fmoc-amino)ethoxy]ethylamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
87556 Aldrich

2-[2-(Fmoc-amino)ethoxy]ethylamine hydrochloride

≥97.0% (HPLC)

Synonym: N-Fmoc-2,2′-oxydiethylamine hydrochloride

  • CAS Number 221352-88-1

  • Empirical Formula (Hill Notation) C19H22N2O3 · HCl

  • Molecular Weight 362.85

  •  MDL number MFCD01321025

  •  PubChem Substance ID 24888912

Purchase

Properties

Related Categories Bifunctional Linkers, Building Blocks, Chemical Biology, Chemical Synthesis, Linkers and Crosslinkers,
InChI Key   ZRXOWZYREIYUHG-UHFFFAOYSA-N
assay   ≥97.0% (HPLC)

Description

Packaging

500 mg in poly tube

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?