• USA Home
  • 88458 - 1-Butyl-4-methylpyridinium hexafluorophosphate

EMAIL THIS PAGE TO A FRIEND
88458 Aldrich

1-Butyl-4-methylpyridinium hexafluorophosphate

≥97.0%

Synonym: 4MBPPF6

Purchase

Properties

Related Categories Chemical Synthesis, Ionic Liquids, Pyridinium, Specialty Synthesis
InChI Key   XBSZSSGZQFAUPF-UHFFFAOYSA-N
assay   ≥97.0%
mp   ~45 °C

Description

Other Notes

Ionic liquid: reaction kinetics of redox reactions

Packaging

5 g in poly bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?