EMAIL THIS PAGE TO A FRIEND
90535 Aldrich

Triethylphosphine

≥97.0%

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Phosphine Compounds, Phosphine Ligands
InChI Key   RXJKFRMDXUJTEX-UHFFFAOYSA-N
assay   ≥97.0%
refractive index   n20/D 1.456(lit.)
bp   127-128 °C(lit.)
density   0.802 g/mL at 20 °C(lit.)

Description

Packaging

ampoule with 10 g

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2845 4.2
WGK Germany 
3
RTECS 
SZ3400000
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

245275 Triethylphosphine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?