• USA Home
  • 90806 - Tributylmethylammonium chloride polymer-bound

EMAIL THIS PAGE TO A FRIEND
90806 Aldrich

Tributylmethylammonium chloride polymer-bound

extent of labeling: ~1.2 mmol/g loading

Synonym: Methyltributylammonium chloride polymer-bound, Tributylammoniomethyl-polystyrene crosslinked with divinylbenzene

Purchase

Properties

Related Categories Chemical Synthesis, Other Supported Reagents, Solid Supported Synthesis, Specialty Synthesis, Supported Reagents More...
extent of labeling   ~1.2 mmol/g loading
matrix   crosslinked with 1% DVB
particle size   200-400 mesh

Description

Other Notes

Immobilized tetraalkylammonium salt with high catalytic activity in PT reactions

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?