• USA Home
  • 91042 - Tricaprylylmethylammonium chloride

EMAIL THIS PAGE TO A FRIEND
91042 Aldrich

Tricaprylylmethylammonium chloride

mixture of C8-C10 C8 is dominant

Synonym: Aliquat® 128, Aliquat® 336 replacement product, Methyltrioctylammonium chloride, Trioctylmethylammonium chloride

Purchase

Properties

Related Categories Ammonium Salts, Biochemicals and Reagents, Cationic Detergents, Chemical Synthesis, Detergents,
description   cationic
InChI Key   XKBGEWXEAPTVCK-UHFFFAOYSA-M
density   0.88 g/mL at 20 °C(lit.)

Description

Application

Quat suitable for nearly all PTC applications without aqueous work-up

Packaging

1 L in glass bottle

250 mL in glass bottle

Legal Information

Aliquat is a registered trademark of BASF

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN2810 - class 6.1 - PG 3 - EHS - Toxic, liquids, organic, n.o.s., HI: all
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?