• USA Home
  • 92079 - 1-(2,4,6-Triisopropylbenzenesulfonyl)-3-nitro-1H-1,2,4-triazole

EMAIL THIS PAGE TO A FRIEND
92079 Aldrich

1-(2,4,6-Triisopropylbenzenesulfonyl)-3-nitro-1H-1,2,4-triazole

≥95.0%

Synonym: TPSNI

  • CAS Number 73118-37-3

  • Empirical Formula (Hill Notation) C17H24N4O4S

  • Molecular Weight 380.46

  •  Beilstein Registry Number 585765

  •  MDL number MFCD00009755

  •  PubChem Substance ID 57653466

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Triazoles
InChI Key   XJBNCASBZVSLGO-UHFFFAOYSA-N
assay   ≥95.0%
mp   128-130 °C(lit.)
  ~130 °C
storage temp.   −20°C

Description

Other Notes

Efficient condensing agent in the triesters synthesis of oligonucleotides

Packaging

1 g in glass bottle

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?