• USA Home
  • 92188 - 2,4,6-Trimethoxyphenylacetic acid

EMAIL THIS PAGE TO A FRIEND
92188 Aldrich

2,4,6-Trimethoxyphenylacetic acid

≥97.0%

  • CAS Number 104397-80-0

  • Empirical Formula (Hill Notation) C11H14O5

  • Molecular Weight 226.23

  •  Beilstein Registry Number 3319447

  •  MDL number MFCD00016821

  •  PubChem Substance ID 57653470

Purchase

Properties

Related Categories Building Blocks, C11 to C12, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   XVJDJIVBWDQELV-UHFFFAOYSA-N
assay   ≥97.0%
mp   182-185 °C

Description

Packaging

1 g in glass bottle

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?