• USA Home
  • 92787 - Ethyl 3-(trimethylsilyl)propionate

EMAIL THIS PAGE TO A FRIEND
92787 Aldrich

Ethyl 3-(trimethylsilyl)propionate

≥95.0%

  • CAS Number 17728-88-0

  • Empirical Formula (Hill Notation) C8H18O2Si

  • Molecular Weight 174.31

  •  Beilstein Registry Number 1759717

  •  MDL number MFCD00010759

  •  PubChem Substance ID 57653493

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   JCANPYOQOFSGFD-UHFFFAOYSA-N
assay   ≥95.0%
refractive index   n20/D 1.420
bp   178-180 °C
density   0.878 g/mL at 20 °C(lit.)

Description

Other Notes

Starting material used for the synthesis of 2-(TMS-methyl)allylic alcohols and similar compounds

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3272 3 / PGIII
WGK Germany 
3
Flash Point(F) 
127.4 °F
Flash Point(C) 
53 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?