• USA Home
  • 93118 - Trityl chloride, polymer-bound

EMAIL THIS PAGE TO A FRIEND
93118 Aldrich

Trityl chloride, polymer-bound

200-400 mesh, extent of labeling: 1.0-2.0 mmol/g loading

Synonym: Chlorotriphenylmethane, polymer-bound, Polystyrene crosslinked with divinylbenzene, α-chlorobenzhydrylated, Triphenylchloromethane, polymer-bound, Triphenylmethyl chloride

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Polymer Supports for SPPS, Resins for Fmoc-/tBu-SPPS,
extent of labeling   1.0-2.0 mmol/g loading
matrix   crosslinked with 1% DVB
particle size   200-400 mesh

Description

Packaging

1 g in poly tube

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?