EMAIL THIS PAGE TO A FRIEND
93925 Aldrich

N-Fmoc-guvacine

≥96.0% (HPLC)

Synonym: 1-Fmoc-1,2,5,6-tetrahydropyridine-3-carboxylic acid

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Other Unnatural Amino Acids & Derivatives, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   YSQFCCXPDNQENP-UHFFFAOYSA-N
assay   ≥96.0% (HPLC)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?