• USA Home
  • 94636 - H-Asn(Trt)-2-ClTrt resin

EMAIL THIS PAGE TO A FRIEND
94636 Aldrich

H-Asn(Trt)-2-ClTrt resin

75-150 μm, extent of labeling: ~0.6 mmol/g loading

Synonym: Nγ-Trityl-L-asparagine 2-chlorotrityl ester polymer-bound

Purchase

Properties

Related Categories 2-Chlorotrityl Resin, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Polymer Supports for SPPS,
extent of labeling   ~0.6 mmol/g loading
particle size   75-150 μm
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

2-Chlorotrityl Resins Preloaded with N-α-Fmoc-Protected Amino Acids

The mild cleavage conditions for highly acid-labile 2-chlorotrityl resin enables the preparation of fully protected peptide fragments for convergent synthesis and selective side-chain derivatization....
Chemfiles Volume 3 Article 4
Keywords: Coupling reactions, Derivatizations

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?