EMAIL THIS PAGE TO A FRIEND
95975 Aldrich

Z-DL-Asp-OH

≥98.0%

Synonym: Z-DL-aspartic acid

  • CAS Number 4515-21-3

  • Empirical Formula (Hill Notation) C12H13NO6

  • Molecular Weight 267.23

  •  Beilstein Registry Number 4701629

  •  MDL number MFCD00063181

  •  PubChem Substance ID 24890227

Purchase

Properties

Related Categories Amino Acids & Derivatives, Aspartic Acid, Chemical Biology, Chemical Synthesis, Peptide Chemistry More...
InChI Key   XYXYXSKSTZAEJW-UHFFFAOYSA-N
assay   ≥98.0%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

A Micro Review of Reversible Addition/Fragmentation Chain Transfer (RAFT) Polymerization

RAFT (Reversible Addition/Fragmentation Chain Transfer) polymerization is a reversible deactivation radical polymerization (RDRP) and one of the more versatile methods for providing living characteri...
Keywords: Biomaterials, Living polymerization, Nanotechnology, Polymerization reactions, Radical polymerization, Reversible addition-fragmentation chain transfer polymerizations, transformation

Concepts and Tools for RAFT Polymerization

RAFT or Reversible Addition/Fragmentation Chain Transfer is a form of living radical polymerization. RAFT polymerization was discovered at CSIRO in 1998.1 It soon became the focus of intensive resear...
Keywords: Polymerization reactions, Radical polymerization, Reversible addition-fragmentation chain transfer polymerizations

Tools for Performing ATRP

Controlled radical polymerization (CRP) techniques have been reviewed in a number of articles1,2 and books,3 and include nitroxide-mediated polymerization (NMP),4-7 reversible addition fragmentation ...
Keywords: Ligands, Materials Science, Polymerization reactions, Purification, Radical polymerization, Reversible addition-fragmentation chain transfer polymerizations, Solution polymerization

Typical Procedures for Polymerizing via RAFT

RAFT (Reversible Addition/Fragmentation Chain Transfer) is a form of living radical polymerization involving conventional free radical polymerization of a substituted monomer in the presence of a sui...
Keywords: Nuclear magnetic resonance spectroscopy, Polymerization reactions, Radical polymerization, Reversible addition-fragmentation chain transfer polymerizations

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?