• USA Home
  • 96022 - (R)-1-Z-2-tert-butyl-3-methyl-4-imidazolidinone

EMAIL THIS PAGE TO A FRIEND
96022 Aldrich

(R)-1-Z-2-tert-butyl-3-methyl-4-imidazolidinone

≥99.0% (N)

  • CAS Number 119906-46-6

  • Empirical Formula (Hill Notation) C16H22N2O3

  • Molecular Weight 290.36

  •  Beilstein Registry Number 5286701

  •  MDL number MFCD00075099

  •  PubChem Substance ID 24890240

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Precursors for Amino Acid Synthesis
InChI Key   TZEQABAXFGVJED-CQSZACIVSA-N
assay   ≥99.0% (N)
optical activity   [α]20/D +10.5±1°, c = 1% in chloroform
mp   83-85 °C
storage temp.   2-8°C

Description

Other Notes

Chiral glycine derivative used for the synthesis of amino acids

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?