EMAIL THIS PAGE TO A FRIEND
96070 Aldrich

Z-Cys(Z)-OH

≥98.0% (HPLC)

Synonym: N,S-Di-Z-L-cysteine

  • CAS Number 57912-35-3

  • Empirical Formula (Hill Notation) C19H19NO6S

  • Molecular Weight 389.42

  •  Beilstein Registry Number 2547850

  •  EC Number 261-013-7

  •  MDL number MFCD00037823

  •  PubChem Substance ID 24890249

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Cysteine/Cystine, Peptide Chemistry More...
InChI Key   PXKPRICKEUGRRR-INIZCTEOSA-N
assay   ≥98.0% (HPLC)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?