EMAIL THIS PAGE TO A FRIEND
96075 Aldrich

Z-dehydro-Ala-OH

≥96.0% (TLC)

Synonym: Z-dehydroalanine

  • CAS Number 39692-63-2

  • Empirical Formula (Hill Notation) C11H11NO4

  • Molecular Weight 221.21

  •  Beilstein Registry Number 2053798

  •  MDL number MFCD00056722

  •  PubChem Substance ID 57653735

Purchase

Properties

Related Categories Alanine Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   QCHJVTWVRPQBBU-UHFFFAOYSA-N
assay   ≥96.0% (TLC)

Description

Packaging

1 g in poly bottle

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?