EMAIL THIS PAGE TO A FRIEND
96253 Aldrich

Boc-Orn(Alloc)-OH

≥97.0%

Synonym: Nα-Boc-Nδ-Alloc-L-ornithine, Nδ-Alloc-Nα-Boc-L-ornithine

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Ornithine Derivatives, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   OSIUYHBPBBOKMJ-JTQLQIEISA-N
assay   ≥97.0%

Description

Packaging

10 g in poly bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?