EMAIL THIS PAGE TO A FRIEND
97005 Aldrich

Z-DL-Phe-OH

≥98.0%

Synonym: Z-DL-phenylalanine

  • CAS Number 3588-57-6

  • Empirical Formula (Hill Notation) C17H17NO4

  • Molecular Weight 299.32

  •  Beilstein Registry Number 3594516

  •  EC Number 222-726-9

  •  MDL number MFCD00063150

  •  PubChem Substance ID 57653801

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylalanine More...
InChI Key   RRONHWAVOYADJL-UHFFFAOYSA-N
assay   ≥98.0%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?