-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
Synonym:
11-
CAS Number 852527-61-8
Empirical Formula (Hill Notation) C11H6F17N3
Molecular Weight 503.16
MDL number MFCD09265211
PubChem Substance ID 24890384
| Related Categories | Aliphatic Azides, Azides, Building Blocks, Chemical Synthesis, Fluorous Synthesis, |
| assay | ≥97.0% |
5, 25 mg in glass btl
Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?