| Related Categories | Analytical/Chromatography, Chromatography, Chromatography, TLC Viewing Cabinets, LPLC/TLC/Paper Chromatography, Labware, |
| W × L × H | 12 in. × 16 in. × 9 in. |
Vinyl-clad aluminum cabinets have separate short- and longwave UV light sources with specular reflectors for maximum intensity and fluorescent contrast. Includes three 8 W light tubes; short- and longwave, and white light for visible illumination. Cabinets have a flexible, contoured eyepiece for UV-absorbing window, color-coded power buttons, push-button wavelength selection, and removable bottom panels for use with transilluminators. UL listed and meets OSHA requirements.
CX is a trademark of Beckman Coulter, Inc.
Spectroline is a registered trademark of Spectronics, Corp.
Customers Also Viewed
8 W, white light