| Related Categories | Eye Protection Equipment, Eyeshields, Labware, Safety Equipment |
| availability | not available in USA and Canada |
A classic, tried and tested design with moulded frame, side shields and available either with abrasion resistant PC lenes for high impact protection or toughened glass lenses for greater chemical resistance. PC lenses BS 2092.2, toughtened glass lenses BS 2092. Spring top case with pocket clip provides protection for these high quality spectacles when they are not in use.